AutoScript TEM Software for Transmission Electron Microscopes
Thermo Scientific™
AutoScript TEM Software for Transmission Electron Microscopes
AutoScript TEM Software offers a Python-based scripting environment for transmission electron microscope operation including automated image acquisition.
Have Questions?
Número de catálogo AUTOSCRIPT-TEM
Precio (MXN)
-
Thermo Scientific AutoScript TEM Software is a scripting environment combining a TEM-specific Python API with the Python editor. Thermo Scientific software packages for electron/ion microscopy, such as Maps Software and Tomography Software for automation and Velox Software for mapping, are great for making collection of standard information easy. However, industrial automation and fundamental research often require advanced techniques for imaging and analysis that cannot be covered in the scope of general purpose software. AutoScript TEM Software provides access to the microscope controls in a Python environment to define your application tailored to the question at hand.
Find additional information here: AutoScript TEM Software
Key Features
Automated electron microscopy and TEM imaging
Integrated IDE
An integrated development environment (IDE) makes it easy to get up and running with AutoScript TEM Software. Object browsing and syntax tools with auto completion are all included to ensure a rich user experience and rapid, consistent scripting framework.
Python
Harness the power of your microscope using the most popular scientific programming language. AutoScript TEM Software is built on Python 3.5 and includes a number of pre-installed libraries for scientific computing, data analysis, data visualization, image processing, and machine learning.
Especificaciones
DescripciónAutoScript TEM Software is a Python-based Application Programming Interface (API) that offers control of Thermo Scientific TEM systems. It opens up the microscope to a world of advanced functions that can be employed for powerful automation.
TipoImaging Data Management Platform