Search
Search
| Catalog Number | Type |
|---|---|
| AY1265X1 | Control Panel Mounting Assembly |
| D0712 | Ultrapure Filter, 1/2 Size |
| D0749 | Filter, 0.22 Micron |
| D0715 | Filter, Oxygen |
| AY1367X1 | Dispense Overflow Cutoff Float |
| AY1403X3 | Pump Interlock Cable |
| D11981 | Remote Dispenser |
| D13661 | Volumetric Remote Dispenser |
| D502113 | Prefilter |
| D502114 | Cartridge, MPS |
| D502115 | Cartridge, Carbon |
| D502137 | Cartridge, TII Filter |
| D502138 | Start-up Kit |
| D7427 | Reverse Osmosis Mixing Valve |
| FL1265X1 | Reverse Osmosis Membrane |
| LMX31 | UV Lamp for Storage Reservoir |