Thermo Scientific™ 2D Tank Liner, ASI™ 26 film
Thermo Scientific™

Thermo Scientific™ 2D Tank Liner, ASI™ 26 film

The 560 L Thermo Scientific™ 2D Tank Liner System without bottom drain is composed of Thermo Scientific™ ASI™ 26 Polyethylene자세히 알아보기
Have Questions?
보기 방식 변경buttonViewtableView
카탈로그 번호용량(미터법)Tubing Style
B100037I
B100037-I으로도 사용됨
200 LNo Tubing
B100051I
B100051-I으로도 사용됨
560 LNo Tubing
B100048I
B100048-I으로도 사용됨
50 LNo Tubing
B100049I340 LNo Tubing
B100038I
B100038-I으로도 사용됨
130 LNo Tubing
B100050I
B100050-I으로도 사용됨
400 LNo Tubing
카탈로그 번호 B100037I
B100037-I으로도 사용됨
제품 가격(KRW)
-
용량(미터법):
200 L
Tubing Style:
No Tubing
The 560 L Thermo Scientific™ 2D Tank Liner System without bottom drain is composed of Thermo Scientific™ ASI™ 26 Polyethylene Film, a durable 5.0 mL film, with dimensions optimized for Thermo Scientific™ drums and commonly-used, industry standard cylindrical tanks. Standard Thermo Scientific tank liners feature an open top for maximum recovery, with drum dolly available to provide mobility.

Features
• Minimizes bioburden due to construction under sterile cGMP conditions
• Eliminates post-use cleaning steps required with reusable containers, helping to reduce cross-contamination risks

Applications
• Removes the need for tank cleaning and helps to reduce cycle times
• Facilitates hydration of powdered media and buffers, and pooling of non-sterile solutions and fluids

사양
치수55.9 cm x 91.4 cm (22 in. x 36 in.)
용도(장비)SV30113
제품라인HyPerforma
수량20 per case
배송 조건Room Temperature
멸균 방법Irradiated
용량(미터법)200 L
Film DescriptionASI 26
물질Polyethylene
Port Detail0 Ports
Tubing StyleNo Tubing
Unit Size200 L
구성 및 보관
Store at room temperature.