Thermo Scientific™
Slide File Storage System
Related applications:
Anatomical Pathology
Save space by storing materials in the stackable Thermo Scientific™ Slide File Storage System. Features a transparent hinged cover and numbered slots for easy identification.
Back to top
Storage:
  • Stores 3 × 1 in.(75 × 26mm) slides
  • Stores up to 100 slides (one per slot), 200 slides (two per slot), or 400 slides (slide tray removed)