Bolt™ Western Pack B (PVDF)
Invitrogen™

Bolt™ Western Pack B (PVDF)

The Bolt™ Western Pack (PVDF) includes two Mini Blot Modules and all of the buffers and pre-cut membrane/filter paper sandwichesRead more
Have Questions?
Catalog NumberDimensions (LxW)
B1000BMini, 8 cm x 8 cm
Catalog number B1000B
Price (EUR)
1.076,88
Online exclusive
1.282,00
Save 205,12
Each
Dimensions (LxW):
Mini, 8 cm x 8 cm
Request bulk or custom format
Price (EUR)
1.076,88
Online exclusive
1.282,00
Save 205,12
Each
The Bolt™ Western Pack (PVDF) includes two Mini Blot Modules and all of the buffers and pre-cut membrane/filter paper sandwiches you will need for the wet transfer of proteins from Bolt™ Bis-Tris Plus gels to PVDF membranes. Each pack contains:
• Mini Blot Modules
• Bolt™ Transfer Buffer (20X)
• Bolt™ Antioxidant
• PVDF pre-cut membranes (0.2 μm)

About the Mini Blot Module
The Mini Blot Module is a wet transfer device used exclusively with the Mini Gel Tank. The tank accommodates one blot module per chamber, or two blot modules total with the side-by-side layout. The Mini Blot Module is compatible with Bolt™ Bis-Tris Plus gels and accommodates one gel per wet transfer. The universal connection and molded gasket make the blot module easy to use, while the inner core of the blot module allows for use of less methanol-based transfer buffer per wet transfer. At the recommended conditions and constant voltage, proteins can be transferred to nitrocellulose or PVDF membranes typically in 30 to 60 minutes.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
For Use With (Equipment)Bolt™
Mode of TransferWet
Quantity≤ 2 Blot Modules/Mini-Gel Tank, 1 Mini-Gel/Blot Module kit
Dimensions (LxW)Mini, 8 cm x 8 cm
MaterialPVDF
Product LineBolt
Unit SizeEach
Contents & Storage
2 Mini Blot Modules
125 mL Bolt™ Transfer Buffer (20X)
15 mL Bolt™ Antioxidant
20-pack PVDF membranes (0.2 μm)

Frequently asked questions (FAQs)

Can I prepare my protein sample with the reducing agent and store it for future use?

DTT is not stable, so it must be added and the reduction performed just prior to loading your samples.

Find additional tips, troubleshooting help, and resources within our Protein Gel 1D Electrophoresis Support Center.

My LDS or SDS sample buffer precipitates when stored at 4 degrees C. Can I warm it up? Can I store it at room temperature?

Precipitation of the LDS or SDS at 4 degrees C is normal. Bring the buffer to room temperature and mix until the LDS/SDS goes into solution. If you do not want to wait for it to dissolve, you can store the sample buffer at room temperature.

Find additional tips, troubleshooting help, and resources within our Protein Gel 1D Electrophoresis Support Center.

What are the storage conditions for Bolt gels?

Similar to NuPAGE gels with storage temperatures of 4 to 25 degrees C.

Find additional tips, troubleshooting help, and resources within our Protein Electrophoresis and Western Blotting Support Center.

What is the shelf life for Bolt gels?

The shelf life for Bolt gels is similar to that for NuPAGE gels: 12 months from the date of shipment.

Find additional tips, troubleshooting help, and resources within our Protein Electrophoresis and Western Blotting Support Center.

How are Bolt gels different than NuPAGE gels?

While they are both Bis-Tris based gels, the chemistries are very different since Bolt gels are optimized for western blotting. Another key difference is the wedge well design of the Bolt gels, which allows larger sample volumes to be loaded.

Find additional tips, troubleshooting help, and resources within our Protein Gel 1D Electrophoresis Support Center.