Thermo Scientific™
Modular Cassette Storage Drawers
 Related applications:
Anatomical Pathology
Permanently store up to 165 embedding rings or 250 cassettes per drawer with Thermo Scientific™ Modular Cassette Storage Drawers.
Error loading your content!
  Catalog number Price ({{currency}}) Availability Qty
{{product.sku}}
also known as {{product.formattedSku}} 
{{product.availability.message}} Add To Cart
{{addItemToList.successMessage}} {{addItemToList.listName}}
Back to top
Description
Features
  • Made from high-impact polystyrene
  • Designed with Interlocking ridges on top and bottom to permit stacking to any convenient height
Includes
  • Identification labels.
Documents
Manuals & protocols