Biotin Alkyne (PEG4 carboxamide-Propargyl Biotin)
Invitrogen™

Biotin Alkyne (PEG4 carboxamide-Propargyl Biotin)

The hapten, biotin azide is reactive with terminal via a copper-catalyzed click reaction. Biotin can be subsequently detected with streptavidin,Read more
Have Questions?
Catalog NumberQuantity
B101851 mg
Catalog number B10185
Price (USD)
700.00
Each
Quantity:
1 mg
Request bulk or custom format
Price (USD)
700.00
Each
The hapten, biotin azide is reactive with terminal via a copper-catalyzed click reaction. Biotin can be subsequently detected with streptavidin, avidin or NeutrAvidin® biotin-binding protein.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Chemical ReactivityAzide
FormatSolid
Label or DyeBiotin
Molecular Weight (g/mol)528.66 Da
Product TypeBiotin Alkyne
Quantity1 mg
Reactive GroupAlkyne
Reactive MoietyAlkyne
Shipping ConditionRoom Temperature
SolubilityDMSO (Dimethylsulfoxide)
ColorNone
Label TypeBiotin and Other Haptens
Product LineMolecular Probes
Unit SizeEach
Contents & Storage
Store at ≤-20°C and keep desiccated.

Citations & References (8)

Citations & References
Abstract
Identification of protein targets of 4-hydroxynonenal using click chemistry for ex vivo biotinylation of azido and alkynyl derivatives.
Authors:Vila A, Tallman KA, Jacobs AT, Liebler DC, Porter NA, Marnett LJ,
Journal:Chem Res Toxicol
PubMed ID:18232660
'Polyunsaturated fatty acids (PUFA) are primary targets of free radical damage during oxidative stress. Diffusible electrophilic alpha,beta-unsaturated aldehydes, such as 4-hydroxynonenal (HNE), have been shown to modify proteins that mediate cell signaling (e.g., IKK and Keap1) and alter gene expression pathways responsible for inducing antioxidant genes, heat shock proteins, and ... More
Direct in-gel fluorescence detection and cellular imaging of O-GlcNAc-modified proteins.
Authors:Clark PM, Dweck JF, Mason DE, Hart CR, Buck SB, Peters EC, Agnew BJ, Hsieh-Wilson LC,
Journal:J Am Chem Soc
PubMed ID:18683930
'We report an advanced chemoenzymatic labeling strategy for direct fluorescence detection of O-GlcNAc proteins in gels that facilitates proteomic studies and greatly extend the reach of existing technologies. These new tools also enable the expression and dynamics of O-GlcNAc modifications to be monitored by imaging in cells and tissues. ... More
Comparative methods for analysis of protein covalent modification by electrophilic quinoids formed from xenobiotics.
Authors:Yu B, Qin Z, Wijewickrama GT, Edirisinghe P, Bolton JL, Thatcher GR,
Journal:Bioconjug Chem
PubMed ID:19301905
'Conjugation of biotin and fluorophore tags is useful for assaying covalent protein modification. Oxidative bioactivation of selective estrogen receptor modulators (SERMs) yields reactive quinoid electrophiles that covalently modify proteins, and bioactivation is associated with carcinogenic and chemopreventive effects. Identification of the protein targets of electrophilic metabolites is of general importance ... More
HACE1 reduces oxidative stress and mutant Huntingtin toxicity by promoting the NRF2 response.
Authors:Rotblat B, Southwell AL, Ehrnhoefer DE, Skotte NH, Metzler M, Franciosi S, Leprivier G, Somasekharan SP, Barokas A, Deng Y, Tang T, Mathers J, Cetinbas N, Daugaard M, Kwok B, Li L, Carnie CJ, Fink D, Nitsch R, Galpin JD, Ahern CA, Melino G, Penninger JM, Hayden MR, Sorensen PH,
Journal:
PubMed ID:24516159
Oxidative stress plays a key role in late onset diseases including cancer and neurodegenerative diseases such as Huntington disease. Therefore, uncovering regulators of the antioxidant stress responses is important for understanding the course of these diseases. Indeed, the nuclear factor erythroid 2-related factor 2 (NRF2), a master regulator of the ... More
O-linked N-acetylglucosamine modification on CCAAT enhancer-binding protein beta: role during adipocyte differentiation.
Authors:Li X, Molina H, Huang H, Zhang YY, Liu M, Qian SW, Slawson C, Dias WB, Pandey A, Hart GW, Lane MD, Tang QQ,
Journal:J Biol Chem
PubMed ID:19478079
CCAAT enhancer-binding protein (C/EBP)beta is a basic leucine zipper transcription factor family member, and can be phosphorylated, acetylated, and sumoylated. C/EBPbeta undergoes sequential phosphorylation during 3T3-L1 adipocyte differentiation. Phosphorylation on Thr(188) by MAPK or cyclin A/cdk2 primes the phosphorylations on Ser(184)/Thr(179) by GSK3beta, and these phosphorylations are required for the ... More