BackDrop™ Background Suppressor for Microplate Assays - FAQs
BackDrop™ Background Suppressor for Microplate Assays - FAQs
View additional product information for BackDrop™ Background Suppressor for Microplate Assays - FAQs (B10511)
6 product FAQs found
How do I quench any fluorescence outside of my live cells and leave my intracellular staining intact?
There are a number of reagents used to quench fluorescence on the outside of live cells or in the media (background fluorescence). Our Backdrop Background Suppressor reagents are a great option to quench blue, green, and red fluorescent dyes, various media components and fluorescent proteins. Another commonly used reagent is the cell impermeant colorimetric dye, Trypan blue.
The BackDrop background suppressor solutions are supposed to reduce background fluorescence. Does that include sample autofluorescence?
No. The BackDrop dyes will quench any fluorescence of a given wavelength in the extracellular media or buffer, but not inside live cells. It will not quench autofluorescence inside live cells unless the dye is engulfed.
Can I use the BackDrop Background Suppressor reagent as the quencher to reach the inner leaflet of the plasma membrane?
We have not tested this. The BackDrop Background Suppressor reagent is cell impermeant, but if the quenching is caused by a FRET interaction and, if the distance between the BackDrop Background Suppressor outside of the cell is within the Ro distance for FRET with the fluorescent dye in the inner leaflet of the membrane, quenching may still occur.
Would the BackDrop Background Suppressor reagent quench Alexa Fluor 647 signal? What is the mechanism of quenching?
Yes, the BackDrop Background Suppressor reagent can be used to quench Alexa Fluor 647 signal. We have not determined the exact mechanism for the quenching but it depends upon the dye. For some dyes, it may be a FRET interaction, for others the BackDrop Background Suppressor absorbs the photons emitted by the fluorescent dye (inner filter effect).
I understand that some media are worse than others for fluorescence imaging. How do I choose?
Most media contain phenol red, which can quench fluorescent dyes in the visible wavelengths. Most media also contain autofluorescent components, such as riboflavin, which can reduce signal-to-background. We offer FluoroBrite DMEM and HEPES-based Live Cell Imaging Solution, which have been optimized for fluorescent imaging. We also offer a number of media without phenol red. But if none of these are reasonable options for your experiment, then we also offer BackDrop Background Suppressor ReadyProbes Reagent, which can be added to quench media autofluorescence.
I am not seeing a strong degree of change in my ion indicator dye in my positive versus negative controls. How can I correct this?
First, make sure you are exciting and detecting the dye in the appropriate wavelengths. Next, try optimizing the dye concentration with your controls, as well as the dye staining time. Our manuals have some guidelines. If you are using a live-cell system, we also recommend washing out any unreacted dye for many of our products to reduce background fluorescence, or adding a background suppressor such as BackDrop Suppressor ReadyProbes Reagent. Check your instrument settings; some plate readers, for instance, may have a means of adjusting the gain setting to get the best signal-to-background. Finally, some dyes are better than others for degree of change upon ion detection. Contact Tech Support by sending an email to techsupport@thermofisher.com if you would like to discuss dye options.