BCECF, AM (2',7'-Bis-(2-Carboxyethyl)-5-(and-6)-Carboxyfluorescein, Acetoxymethyl Ester)
Invitrogen™

BCECF, AM (2',7'-Bis-(2-Carboxyethyl)-5-(and-6)-Carboxyfluorescein, Acetoxymethyl Ester)

With a pKa of ∼6.98, the cell-permeant, dual-excitation ratiometric pH indicator BCECF, AM is ideal for measuring changes in the자세히 알아보기
Have Questions?
보기 방식 변경buttonViewtableView
카탈로그 번호수량
B117020 x 50 μg
B11501 mg
카탈로그 번호 B1170
제품 가격(KRW)
468,000
온라인 행사
Ends: 31-Dec-2026
534,000
할인액 66,000 (12%)
Each
수량:
20 x 50 μg
제품 가격(KRW)
468,000
온라인 행사
Ends: 31-Dec-2026
534,000
할인액 66,000 (12%)
Each
With a pKa of ∼6.98, the cell-permeant, dual-excitation ratiometric pH indicator BCECF, AM is ideal for measuring changes in the cytosolic pH of most cells. Intracellular pH measurements with BCECF are made by determining the pH-dependent ratio of emission intensity (detected at 535 nm) when the dye is excited at ∼490 nm versus the emission intensity when excited at its isobestic point of ∼440 nm.
For Research Use Only. Not for use in diagnostic procedures.
사양
검출 방법Fluorescence
염료 유형pH Indicator
수량20 x 50 μg
배송 조건Room Temperature
용도 (장비)Microplate Reader
제품 유형pH indicator
Unit SizeEach
구성 및 보관
Store in freezer -5°C to -30°C.

자주 묻는 질문(FAQ)

Why don't I see a significant change in signal for my live-cell fluorescent indicator dye?

Regardless of the type of live-cell indicator dye (e.g., calcium indicators, pH indicator, metal ion indicators), make sure there is no serum during the loading step, which can prematurely cleave dyes with AM esters and bind dyes non-specifically. Always optimize the dye concentration and staining time with a positive control before you run your test samples, to give the best signal-to-background. Always run a positive control with a buffer containing free ions of known concentration and an ionophore to open pores to those ions (for instance, for calcium indicators like Fluo-4 AM, this would include a buffer with added calcium combined with calcimycin, or for pH indicators, buffers of different pHs combined with nigericin). Reactive oxygen indicators, such as CellROX Green or H2DCFDA would require a cellular reactive oxygen species (ROS) stimulant as a positive control, such as menadione. Finally, make sure your imaging system has a sensitive detector. Plate readers, for instance, have much lower detector efficiency over background, compared to microscopy or flow cytometry.

Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.

인용 및 참조 문헌 (1251)

인용 및 참조 문헌
Abstract
Nitric oxide mediates natural polyphenol-induced Bcl-2 down-regulation and activation of cell death in metastatic B16 melanoma.
Authors:Ferrer P,Asensi M,Priego S,Benlloch M,Mena S,Ortega A,Obrador E,Esteve JM,Estrela JM
Journal:The Journal of biological chemistry
PubMed ID:17135264
Authors:
Journal:
PubMed ID:8978825
Platelet-mediated lymphocyte delivery to high endothelial venules.
Authors:Diacovo TG,Puri KD,Warnock RA,Springer TA,von Andrian UH
Journal:Science (New York, N.Y.)
PubMed ID:8662511
Molecular cloning, chromosomal localization, tissue distribution, and functional expression of the human pancreatic sodium bicarbonate cotransporter.
Authors:Abuladze N, Lee I, Newman D, Hwang J, Boorer K, Pushkin A, Kurtz I
Journal:J Biol Chem
PubMed ID:9651366
We report the cloning, sequence analysis, tissue distribution, functional expression, and chromosomal localization of the human pancreatic sodium bicarbonate cotransport protein (pancreatic NBC (pNBC)). The transporter was identified by searching the human expressed sequence tag data base. An I.M.A.G.E. clone W39298 was identified, and a polymerase chain reaction probe was ... More
Transforming growth factor beta 1 increases the number of apoptotic bodies and decreases intracellular pH in isolated periportal and perivenular rat hepatocytes.
Authors:Benedetti A, Di Sario A, Svegliati Baroni G, Jezequel AM
Journal:Hepatology
PubMed ID:7590668
Transforming growth factor beta 1 (TGF beta 1) is involved in promoting cell death by apoptosis in the liver, whereas the activation of Na+/H+ exchanger has been related to cell proliferation. The aim of this study was to gain information on the effects of TGF beta 1 on intracellular pH ... More