BODIPY® 650⁄665 Phalloidin - FAQs
BODIPY® 650⁄665 Phalloidin - FAQs
View additional product information for BODIPY® 650⁄665 Phalloidin - FAQs (B12382)
1 product FAQs found
I'm trying to label my paraffin sections for F-actin with a phalloidin conjugate, but I'm not seeing any signal. Why?
When cells and tissues are treated with solvents such as xylene or acetone (for example during deparaffinization of tissue sections), it affects the F-actin in a way that prevents phalloidins from binding. Phalloidin may be used with cryosections, which are not typically washed with organic solvents, or anti-actin antibodies may be used.