Co-factors
Thermo Scientific™

Co-factors

Thermo Scientific enzymes for in vitro transcription are designed for efficient and reliable RNA synthesis. These enzymes cover the entire synthesis workflow from template preparation to post-transcriptional modifications, ensuring optimal performance and high yields of synthesized RNA.
Have Questions?
Cambiar vistabuttonViewtableView
Número de catálogoCantidadTipo de productoConcentración
B13880C800 μLDonante del grupo metilo5 mM
C24341250 μLCofactor de la polimerasa100 mM
F-5101250 μLCofactor de la polimerasa50 mM
Número de catálogo B13880C
Precio (MXN)
-
Cantidad:
800 μL
Tipo de producto:
Donante del grupo metilo
Concentración:
5 mM
Pedido a granel o personalizado

Thermo Scientific enzymes for in vitro transcription are designed for efficient and reliable RNA synthesis. These enzymes cover the entire synthesis workflow from template preparation to post-transcriptional modifications, ensuring optimal performance and high yields of synthesized RNA.

High quality RNA synthesized by in vitro transcription is essential for RNA research and this versatile technique can be used in a variety of applications including the introduction of modified nucleotides into RNA transcripts for enhanced translational capacity and reduced immune activity of the cells.

Thermo Scientific enzymes for in vitro transcription are designed for efficient and reliable RNA synthesis. These enzymes cover the entire synthesis workflow from template preparation to post-transcriptional modifications, ensuring optimal performance and high yields of synthesized RNA.

High quality RNA synthesized by in vitro transcription is essential for RNA research and this versatile technique can be used in a variety of applications including the introduction of modified nucleotides into RNA transcripts for enhanced translational capacity and reduced immune activity of the cells.

Features

  • Convenient stand-alone enzymes and buffers for workflow optimization
  • No animal-origin components in final product composition
  • Reliable and reproducible results with rigorous quality testing
  • Scalable and available as OEM products

Applications

  • CRISPR/Cas-based genome editing
  • Gene expression
  • Generation of pluripotent cells

Para uso exclusivo en investigación. No apto para uso en procedimientos diagnósticos.

Especificaciones
Nombre del producto químico o materialS-Adenosyl-L- Methionine (SAM)
N.º de reacciones100 Reactions
Tipo de productoDonante del grupo metilo
Cantidad800 μL
Concentración5 mM
FormularioLíquido
FormatoVial
Unit SizeEach
Contenido y almacenamiento
5 mM SAM, 800 μL

Store at -20°C.