Product Image
Thermo Scientific™

Taq Buffers (10X)

Thermo Scientific 10X Taq Buffers are ready-to-use buffers for PCR using Taq DNA Polymerases (both recombinant and native). Available in different compositions and also without detergents.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberChemical Name or Material
B16Taq Buffer with KCl and MgCl2
B55Taq Buffer without Detergent
B38Taq Buffer with KCl
B33Taq Buffer with (NH4)2SO4
B34Taq Buffer with (NH4)2SO4 and MgCl2
Catalog number B16
Price (USD)
29.62
Each
Chemical Name or Material:
Taq Buffer with KCl and MgCl2
Request bulk or custom format
Price (USD)
29.62
Each
Thermo Scientific 10X Taq Buffers are ready-to-use buffers for PCR using Taq DNA polymerases (both recombinant and native). They are available in different compositions and also without detergents.

10X Taq Buffer without Detergent

  • 100 mM Tris-HCl (pH 8.8 at 25°C)
  • 500 mM KCl

10X Taq Buffer with KCl and 15 mM MgCl2

  • 100 mM Tris-HCl (pH 8.8 at 25°C)
  • 500 mM KCl
  • 15 mM MgCl2
  • Detergent

10X Taq Buffer with (NH4)2SO4

  • 750 mM Tris-HCl (pH 8.8 at 25°C)
  • 200 mM (NH4)2SO4
  • 0.1% (v/v) Tween 20

10X Taq Buffer with (NH4)2SO4 and 20 mM MgCl2

  • 750 mM Tris-HCl (pH 8.8 at 25°C)
  • 200 mM (NH4)2SO4
  • 0.1% (v/v) Tween 20
  • 20 mM MgCl2

10X Taq Buffer with KCl

  • 100 mM Tris-HCl (pH 8.8 at 25°C)
  • 500 mM KCl
  • Detergent
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Chemical Name or MaterialTaq Buffer with KCl and MgCl2
Includes• 10X Taq Buffer with KCl and 15 mM MgCl2, 4 x 1.25 mL

Store at -20°C.
Concentration10X
Physical FormLiquid
Quantity4 x 1.25 mL
Unit SizeEach