Thermo Scientific™
Plastic Slide Boxes
 Related applications:
Anatomical Pathology
Store a lot of slides in a little space with durable Thermo Scientific™ Plastic Slide Boxes.
Error loading your content!
  Catalog number Price ({{currency}}) Availability Qty
{{product.sku}}
also known as {{product.formattedSku}} 
{{product.availability.message}} Add To Cart
{{addItemToList.successMessage}} {{addItemToList.listName}}
Back to top
Description
  • Durable ABS plastic construction and internal cork lining safeguards slides
  • Store and protect 25, 50 or 100 slides
  • Index card fits in lid to record slide titles
  • Numbered slots on box correspond to lid index to maintain organization
  • Lids separate from 25- and 50-slide boxes for easy storage and retrieval
  • Available in red, white or blue for color-coding or aesthetic preference
  • 50-slide box opens and shuts using a plastic latch; 100-slide box has a hinged lid with a metal clasp
Dimensions (closed)
25-Slide Box: 5.5 × 3.375 × 1.25 in. (9.5 × 8.3 × 3.8cm)
50-Slide Box: 8.25 in. × 3.375 in. × 1.25 in. (21 × 8.5 × 3cm)
100-Slide Box: 8.25 x 6.375 x 1.25 in. (21 x 16 x 3cm)
Please Note: Availability may vary by country.
Documents
Manuals & protocols