Owl™ EasyCast™ B1A Mini Gel and A2-OK Multi Gel Electrophoresis System Combs
Actual product may vary
Owl™ EasyCast™ B1A Mini Gel and A2-OK Multi Gel Electrophoresis System Combs
Owl™ EasyCast™ B1A Mini Gel and A2-OK Multi Gel Electrophoresis System Combs
Thermo Scientific™
Owl™ EasyCast™ B1A Mini Gel and A2-OK Multi Gel Electrophoresis System Combs
Eliminate comb assembly with this assortment of heavy-duty, one-piece combs.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberTypeTeeth CountThickness (Metric) Teeth
B1A-PREPPrep Comb21.5 mm
B1A-5CComb51 mm
B1A-5DComb51.5 mm
B1A-6Double-sided Comb61.0/1.5 mm
B1A-8Double-sided Comb81.0/1.5 mm
Catalog number B1A-PREP
Price (USD)
80.28
Online exclusive
89.20
Save 8.92
Each
Type:
Prep Comb
Teeth Count:
2
Thickness (Metric) Teeth:
1.5 mm
Price (USD)
80.28
Online exclusive
89.20
Save 8.92
Each
Eliminate comb assembly with Thermo Scientific™ Owl™ EasyCast™ B1A Mini Gel and A2-OK Multi Gel Electrophoresis System Combs. This assortment of heavy-duty, one-piece combs is for use with the EasyCast B1A Mini Gel and A2-OK Multi Gel Electrophoresis System.
Ordering Information:
Customized combs are available (order SKU#: XCM). Please contact your customer service or sales representative for further details.
Specifications
No. per Pack96
Teeth Count2
Thickness (Metric) Teeth1.5 mm
For Use With (Equipment)Owl™ EasyCast B1A Mini Gel and A2-OK Multi Gel Electrophoresis System
Gel SizeMini
Product LineOwl, EasyCast
TypePrep Comb
Unit SizeEach
Frequently asked questions (FAQs)
Are Bio-Rad power supplies compatible with Owl Electrophoresis systems?
The Owl Electrophoresis Systems are supplied with standard power leads, but we have not tested compatibility of the Owl systems with the Bio-Rad power supplies. For best performance, we recommend using one of our Owl or PowerEase power supplies for the Owl Electrophoresis Systems.