4-(1-Piperazinyl)phenol, 95%
4-(1-Piperazinyl)phenol, 95%
4-(1-Piperazinyl)phenol, 95%
4-(1-Piperazinyl)phenol, 95%
Thermo Scientific Chemicals

4-(1-Piperazinyl)phenol, 95%

CAS: 56621-48-8 | C10H14N2O | 178.24 g/mol
Have Questions?
變更視圖buttonViewtableView
Quantity:
10 g
50 g
250 g
產品號碼 B20122.09
亦稱為 B20122-09
價格 (TWD)
-
申請報價
Quantity:
10 g
Request bulk or custom format
化學識別
CAS56621-48-8
IUPAC Name4-(piperazin-1-yl)phenol
Molecular FormulaC10H14N2O
InChI KeyGPEOAEVZTOQXLG-UHFFFAOYSA-N
SMILESOC1=CC=C(C=C1)N1CCNCC1
檢視更多
規格Specification Sheet規格表
FormPowder
Assay (Non-aqueous acid-base Titration)≥94.0 to ≤106.0%
Appearance (Color)Pale brown to brown or pink
Assay (HPLC)≥94.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only