[2.2]Paracyclophane, 99%
[2.2]Paracyclophane, 99%
[2.2]Paracyclophane, 99%
Thermo Scientific Chemicals

[2.2]Paracyclophane, 99%

CAS: 1633-22-3 | C16H16 | 208.304 g/mol
Have Questions?
變更視圖buttonViewtableView
Quantity:
1 g
5 g
產品號碼 B20136.03
亦稱為 B20136-03
價格 (TWD)
-
Quantity:
1 g
Request bulk or custom format
化學識別
CAS1633-22-3
IUPAC Nametricyclo[8.2.2.2⁴,⁷]hexadeca-1(12),4,6,10,13,15-hexaene
Molecular FormulaC16H16
InChI KeyOOLUVSIJOMLOCB-UHFFFAOYSA-N
SMILESC1CC2=CC=C(CCC3=CC=C1C=C3)C=C2
檢視更多
規格Specification Sheet規格表
Assay (HPLC)≥98.5%
FormCrystals or powder or crystalline powder
Appearance (Color)White to pale yellow
2,2]-Paracyclophane is the raw material for the synthesis of parylene which is widely used in microelectronic integrated circuit.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,2]-Paracyclophane is the raw material for the synthesis of parylene which is widely used in microelectronic integrated circuit.

Solubility
Insoluble in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
RUO – Research Use Only

General References:

  1. Susan E. Gibson and Jamie D. Knight. [2.2]Paracyclophane derivatives in asymmetric catalysis.ACS Med Chem Lett.2014,5(4), 434-439.
  2. Stefan Dahmen and Stefan Bräse. The Asymmetric Dialkylzinc Addition to Imines Catalyzed by [2.2]Paracyclophane-Based N,O-Ligands.J. Am. Chem. Soc.,2002124(21), 5940-5941.