3-Nitro-o-phenylenediamine, 98%
3-Nitro-o-phenylenediamine, 98%
3-Nitro-o-phenylenediamine, 98%
Thermo Scientific Chemicals

3-Nitro-o-phenylenediamine, 98%

CAS: 3694-52-8 | C6H7N3O2 | 153.141 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
Catalog number B20201.03
also known as B20201-03
Price (EUR)
54,70
Each
Quantity:
1 g
Request bulk or custom format
Price (EUR)
54,70
Each
Chemical Identifiers
CAS3694-52-8
IUPAC Name3-nitrobenzene-1,2-diamine
Molecular FormulaC6H7N3O2
InChI KeyIOCXBXZBNOYTLQ-UHFFFAOYSA-N
SMILESNC1=CC=CC(=C1N)[N+]([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
FormCrystals or powder or crystalline powder
Appearance (Color)Dark red
Melting Point (clear melt)157.0-164.0?C
Assay (GC)≥97.5%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only