1,3-Di-tert-butylbenzene, 99%
1,3-Di-tert-butylbenzene, 99%
1,3-Di-tert-butylbenzene, 99%
Thermo Scientific Chemicals

1,3-Di-tert-butylbenzene, 99%

CAS: 1014-60-4 | C14H22 | 190.33 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
Catalog number B20290.03
also known as B20290-03
Price (USD)
78.65
Special offer
Online exclusive
Ends: 30-Jun-2026
92.90
Save 14.25 (15%)
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
78.65
Special offer
Online exclusive
Ends: 30-Jun-2026
92.90
Save 14.25 (15%)
Each
Chemical Identifiers
CAS1014-60-4
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
FormLiquid
Refractive Index1.4860-1.4890 @ 20?C
Assay (GC)≥98.5%
1,3-Di-tert-butylbenzene is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,3-Di-tert-butylbenzene is used as a pharmaceutical intermediate.

Solubility
Insoluble in water.

Notes
Store in cool, dry place in a well sealed container. Store away from oxidizing agents.
RUO – Research Use Only

General References:

  1. J T Gerig. Solute-solvent interactions probed by intermolecular NOEs. Journal of Organic Chemistry. 2003, 68 (13), 5244-5248.
  2. James Ray Cozort; James F. OutlawJr., Adrian Hawkins; Samuel Siegel. Dienes as possible intermediates in the catalytic hydrogenation of aromatic hydrocarbons. 2. Dienes derived from 1,3-di-tert-butylbenzene and a rhodium catalyst. J. Org. Chem. 1983, 48 (23), 4190-4197.