Thermo Scientific Chemicals

Sodium 1-hexadecanesulfonate, 97%, Thermo Scientific Chemicals

Catalog number: B20382.22
100 g, Each
Thermo Scientific Chemicals

Sodium 1-hexadecanesulfonate, 97%, Thermo Scientific Chemicals

Catalog number: B20382.22
100 g, Each
Quantity
Catalog number: B20382.22
also known as B20382-22
Price (EUR)
Quantity
-

Chemical Identifiers

CAS
15015-81-3
IUPAC Name
sodium hexadecane-1-sulfonate
Molecular Formula
C16H33NaO3S
InChI Key
PNGBYKXZVCIZRN-UHFFFAOYSA-M
SMILES
[Na+].CCCCCCCCCCCCCCCCS([O-])(=O)=O
Assay (ex Sulphated Ash)
≥96.0 to ≤104.0% (dry wt. basis)
Appearance (Color)
White
Form
Powder
Sulfated Ash
20.76-22.49%
Water Content (Karl Fischer Titration)
≤3.0%

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text