Thermo Scientific Chemicals

3-Ethylcyclopentanone, 99%, Thermo Scientific Chemicals

Catalog number: B20432.14
25 g, Each
Thermo Scientific Chemicals

3-Ethylcyclopentanone, 99%, Thermo Scientific Chemicals

Catalog number: B20432.14
25 g, Each
Quantity
Catalog number: B20432.14
also known as B20432-14
Price (USD)
Price: 803.00
Online price: 694.65
Your price:
Quantity
-

Chemical Identifiers

CAS
10264-55-8
IUPAC Name
3-ethylcyclopentan-1-one
Molecular Formula
C7H12O
InChI Key
XERALSLWOPMNRJ-UHFFFAOYNA-N
SMILES
CCC1CCC(=O)C1
Assay (GC)
≥98.5%
Form
Liquid
Refractive Index
1.4375-1.4405 @ 20?C
Appearance (Color)
Clear colorless to pale yellow

Description

3-Ethylcyclopentanone, is used as a chemical reagent, pharmaceutical reagent and pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Ethylcyclopentanone, is used as a chemical reagent, pharmaceutical reagent and pharmaceutical intermediate.

Solubility
Soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text