(1S)-(-)-Camphorsulfonylimine, 98+%
(1S)-(-)-Camphorsulfonylimine, 98+%
(1S)-(-)-Camphorsulfonylimine, 98+%
Thermo Scientific Chemicals

(1S)-(-)-Camphorsulfonylimine, 98+%

CAS: 60886-80-8 | C10H15NO2S | 213.30 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
Catalog number B20823.14
also known as B20823-14
Price (EUR)
398,00
Each
Quantity:
25 g
Request bulk or custom format
Price (EUR)
398,00
Each
Chemical Identifiers
CAS60886-80-8
IUPAC Name10,10-dimethyl-3λ⁶-thia-4-azatricyclo[5.2.1.0¹,⁵]dec-4-ene-3,3-dione
Molecular FormulaC10H15NO2S
InChI KeyZAHOEBNYVSWBBW-UHFFFAOYNA-N
SMILESCC1(C)C2CCC11CS(=O)(=O)N=C1C2
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to pale cream
FormCrystals or powder or crystalline powder
Assay (GC)≥96.0%
Melting Point (clear melt)225.0-231.0°C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only