Methyl cyclopropanecarboxylate, 98%
Methyl cyclopropanecarboxylate, 98%
Methyl cyclopropanecarboxylate, 98%
Methyl cyclopropanecarboxylate, 98%
Thermo Scientific Chemicals

Methyl cyclopropanecarboxylate, 98%

CAS: 2868-37-3 | C5H8O2 | 100.117 g/mol
製品番号(カタログ番号) B20853.14
または、製品番号B20853-14
価格(JPY)
-
見積もりを依頼する
数量:
25 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS2868-37-3
IUPAC Namemethyl cyclopropanecarboxylate
Molecular FormulaC5H8O2
InChI KeyPKAHQJNJPDVTDP-UHFFFAOYSA-N
SMILESCOC(=O)C1CC1
さらに表示
Refractive Index1.4165-1.4205 @ 20?C
Appearance (Color)Clear colorless to pale yellow
Assay (GC)≥97.5%
FormLiquid
It is primarily used an intermediate in the manufacture of pharmaceutical and agrochemical products. Methyl cyclopropane carboxylate is used as a reagent in the synthesis of N’-thiazole and N-pyrazole based inhibitors of MAP kinase p38α.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is primarily used an intermediate in the manufacture of pharmaceutical and agrochemical products. Methyl cyclopropane carboxylate is used as a reagent in the synthesis of N’-thiazole and N-pyrazole based inhibitors of MAP kinase p38α.

Solubility
Not miscible or difficult to mix in water.

Notes
Store away from oxidizing agents and ignition sources. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Protect from electrostatic charges.
RUO – Research Use Only

General References:

  1. Matthäus Getlik, et al. Structure-based design, synthesis and biological evaluation of N-pyrazole, N'-thiazole urea inhibitors of MAP kinase p38α.Eur. J. Med. Chem.2012481-15.
  2. Roseann Kroeker Sachs, et al. 3-Carbomethoxycyclopropen-3-yl Anion. Formation and Characterization of an Antiaromatic Ion. J. Am. Chem. Soc.,1994,116(2), 783-784.