1-Phenylcyclohexanol, 97%
1-Phenylcyclohexanol, 97%
1-Phenylcyclohexanol, 97%
Thermo Scientific Chemicals

1-Phenylcyclohexanol, 97%

CAS: 1589-60-2 | C12H16O | 176.259 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
Catalog number B20877.09
also known as B20877-09
Price (EUR)
51,90
Each
Quantity:
10 g
Request bulk or custom format
Price (EUR)
51,90
Each
Chemical Identifiers
CAS1589-60-2
IUPAC Name1-phenylcyclohexan-1-ol
Molecular FormulaC12H16O
InChI KeyDTTDXHDYTWQDCS-UHFFFAOYSA-N
SMILESOC1(CCCCC1)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
FormCrystals or powder or crystalline powder or fused solid
Assay (GC)≥96.0%
Appearance (Color)White to pale cream to pale yellow
Melting Point (clear melt)55.0-67.0?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only