A substrate used in a microwave-assisted, palladium-catalyzed arylation of acetone as its TMS enol ether.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
A substrate used in a microwave-assisted, palladium-catalyzed arylation of acetone as its TMS enol ether.
Solubility
Slightly soluble in water.
Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
RUO – Research Use Only
General References:
- Wayne J. Thompson, et al. A general synthesis of 5-arylnicotinates.J. Org. Chem.,1984,49(26), 5237-5243.
- Masahiko Iyoda, et al. Homocoupling of Aryl Halides Using Nickel(II) Complex and Zinc in the Presence of Et4NI. An Efficient Method for the Synthesis of Biaryls and Bipyridines.Bull. Chem. Soc. Jpn.,1990,63(1), 80-87.