3-Methoxyphenylacetone is employed in the preparation of optically active cyanohydrins. It is also used in the synthesis of central nervous system (CNS) active compounds. Further, it plays an important role in the preparation of 4-amino-3-methoxypropiophenone.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
3-Methoxyphenylacetone is employed in the preparation of optically active cyanohydrins. It is also used in the synthesis of central nervous system (CNS) active compounds. Further, it plays an important role in the preparation of 4-amino-3-methoxypropiophenone.
Solubility
Miscible with dimethyl sulfoxide.
Notes
Incompatible with strong oxidizing agents, strong acids and bases.
RUO – Research Use Only
General References:
- Vera, W. J.; Banerjee, A. K. First synthesis of 3-methoxy-4-aminopropiophenone. J. Chem. Res. 2012, 36 (9), 542-543.
- Błachut, D.; Danikiewicz, W.; Olejnik, M.; Czarnocki, Z. Electron ionization mass spectrometry as a tool for the investigation of the ortho effect in fragmentation of some Schiff bases derived from amphetamine analogs. J. Mass Spectrom. 2004, 39 (8), 966-972.