Thermo Scientific Chemicals

4-Carboxybenzeneboronic acid, 97%, Thermo Scientific Chemicals

Catalog number: B20954.14
25 g, Each
Thermo Scientific Chemicals

4-Carboxybenzeneboronic acid, 97%, Thermo Scientific Chemicals

Catalog number: B20954.14
25 g, Each
Quantity
Catalog number: B20954.14
also known as B20954-14
Price (USD)
Price: 592.00
Online price: 506.65
Your price:
Quantity
-

Chemical Identifiers

CAS
14047-29-1
IUPAC Name
4-(dihydroxyboranyl)benzoic acid
Molecular Formula
C7H7BO4
InChI Key
SIAVMDKGVRXFAX-UHFFFAOYSA-N
SMILES
OB(O)C1=CC=C(C=C1)C(O)=O
Appearance (Color)
White to pale cream
Assay (HPLC)
≥96.0%
Assay (Aqueous acid-base Titration)
≥96.0%
Form
Powder or lumps or granules
Identification (FTIR)
Conforms

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text