Thermo Scientific Chemicals

Perfluoro-1-iodohexane, 98+%, stab. with copper, Thermo Scientific Chemicals

Catalog number: B20958.30
250 g, Each
Thermo Scientific Chemicals

Perfluoro-1-iodohexane, 98+%, stab. with copper, Thermo Scientific Chemicals

Catalog number: B20958.30
250 g, Each
Quantity
Catalog number: B20958.30
also known as B20958-30
Price (USD)
Price: 650.00
Online price: 552.65
Your price:
Quantity
-

Chemical Identifiers

CAS
355-43-1
IUPAC Name
1,1,1,2,2,3,3,4,4,5,5,6,6-tridecafluoro-6-iodohexane
Molecular Formula
C6F13I
InChI Key
BULLJMKUVKYZDJ-UHFFFAOYSA-N
SMILES
FC(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)I
Appearance (Color)
Clear colorless to purple
Form
Liquid
Assay (GC)
>98.0%
Comment
Stabilised with Copper powder
Refractive Index
1.3270-1.3310 @ 20?C

Description

It is used as an initiator in the presence of fac-[Ir(ppy) 3 for the photoredox catalyst activated CRP of methacrylates in visible light-induced controlled radical polymerization of methacrylates with perfluoroalkyl iodide.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as an initiator in the presence of fac-[Ir(ppy) 3 for the photoredox catalyst activated CRP of methacrylates in visible light-induced controlled radical polymerization of methacrylates with perfluoroalkyl iodide.

Solubility
Not miscible or difficult to mix in water.

Notes
Light sensitive. Store away from light and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text