2-(Trimethylsilyl)ethanol, 98+%
2-(Trimethylsilyl)ethanol, 98+%
2-(Trimethylsilyl)ethanol, 98+%
Thermo Scientific Chemicals

2-(Trimethylsilyl)ethanol, 98+%

CAS: 2916-68-9 | C5H14OSi | 118.251 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Catalog number B20970.06
also known as B20970-06
Price (USD)
92.20
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
92.20
Each
Chemical Identifiers
CAS2916-68-9
IUPAC Name2-(trimethylsilyl)ethan-1-ol
Molecular FormulaC5H14OSi
InChI KeyZNGINKJHQQQORD-UHFFFAOYSA-N
SMILESC[Si](C)(C)CCO
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
Assay (GC)≥98.0%
Identification (FTIR)Conforms
Refractive Index1.4230-1.4270 @ 20?C
FormLiquid
2-(Trimethylsilyl)ethanol is used as a protecting reagent for carboxyl, phosphoryl, hydroxyl and amino group in organic synthesis. It is used as a precursor to prepare trimethyl(2-phenoxyethyl)silanes by reacting with aromatic fluoride. It is also used in the synthesis of teoc-protected amines by using the corresponding isocyanates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-(Trimethylsilyl)ethanol is used as a protecting reagent for carboxyl, phosphoryl, hydroxyl and amino group in organic synthesis. It is used as a precursor to prepare trimethyl(2-phenoxyethyl)silanes by reacting with aromatic fluoride. It is also used in the synthesis of teoc-protected amines by using the corresponding isocyanates.

Solubility
Miscible with water.

Notes
Keep away from sources of ignition. Keep container tightly closed in a dry and well-ventilated place.
RUO – Research Use Only

General References:

  1. Carboxyl groups can be protected as 2-(trimethylsilyl)ethyl (TMSE) esters by carbodiimide coupling: Helv. Chim. Acta, 60, 2711 (1977); J. Org. Chem., 62, 93 (1997), from the acid chloride in pyridine: Helv. Chim. Acta, 60, 3039 (1977), or refluxing the acid in THF in the presence of TMS chloride: J. Am. Chem. Soc., 106, 3030 (1984). The TMSE protecting group is useful in peptide synthesis. Cleavage is readily effected with TBAF in DMF or DMSO: Helv. Chim. Acta, 60, 2711 (1977); Bioorg. Med. Chem. Lett., 3, 437 (1993).
  2. Qu, H.; Sun, W.; Sun, T.; Sollogoub, M.; Zhang, Y. Synthesis and characterization of four novel 2-(trimethylsilyl)ethyl glycosides. Res. Chem. Intermed. 2015, 41 (2), 1107-1113.
  3. Sano, S.; Sumiyoshi, H.; Handa, A.; Tokizane, R.; Nakao, M. A novel synthetic approach to glycerophospholipids via Horner-Wadsworth-Emmons reaction of mixed phosphonoacetate. Tetrahedron Lett. 2015, 56 (32), 4686-4688.