1,12-Dibromododecane, 98%
1,12-Dibromododecane, 98%
1,12-Dibromododecane, 98%
1,12-Dibromododecane, 98%
Thermo Scientific Chemicals

1,12-Dibromododecane, 98%

CAS: 3344-70-5 | C12H24Br2 | 328.132 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Catalog number B21254.22
also known as B21254-22
Price (TWD)
-
Request A Quote
Quantity:
100 g
Request bulk or custom format
Chemical Identifiers
CAS3344-70-5
IUPAC Name1,12-dibromododecane
Molecular FormulaC12H24Br2
InChI KeyZJJATABWMGVVRZ-UHFFFAOYSA-N
SMILESBrCCCCCCCCCCCCBr
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to cream to pale brown
FormCrystals or powder or crystalline powder
Assay (GC)≥97.5%
Identification (FTIR)Conforms
Melting Point (clear melt)36.0-43.0?C
1,12-Dibromododecane, is used as an organic chemical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,12-Dibromododecane, is used as an organic chemical intermediate.

Solubility
Soluble in hot methanol- almost transparency.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions.Stable. Combustible. Incompatible with strong oxidizing agents.
RUO – Research Use Only

General References:

  1. R Zana.; M Benrraou.; R Rueff. Alkanediyl-.alpha.,.μ.-bis(dimethylalkylammonium bromide) surfactants. 1. Effect of the spacer chain length on the critical micelle concentration and micelle ionization degree. Langmuir. 19917 (6), 1072-1075.