Thermo Scientific Chemicals

5-Iodosalicylic acid, 97%, Thermo Scientific Chemicals

Catalog number: B21291.36
500 g, Each
Thermo Scientific Chemicals

5-Iodosalicylic acid, 97%, Thermo Scientific Chemicals

Catalog number: B21291.36
500 g, Each
Quantity
Catalog number: B21291.36
also known as B21291-36
Price (TWD)

Chemical Identifiers

CAS
119-30-2
IUPAC Name
2-hydroxy-5-iodobenzoic acid
Molecular Formula
C7H5IO3
InChI Key
SWDNKOFGNPGRPI-UHFFFAOYSA-N
SMILES
OC(=O)C1=CC(I)=CC=C1O
Appearance (Color)
White to cream to pale yellow
Form
Crystals or powder or crystalline powder
Assay (Aqueous acid-base Titration)
≥96.0 to ≤104.0%
Assay (Silylated GC)
≥96.0%
Identification (FTIR)
Conforms

Description

2-hydroxy-5-iodobenzoic acid is a reagent in various organic chemical reactions leading to pharmaceutical compounds. It is involved in the synthesis of labeled benzamide analogs for tumor imaging as well as the synthesis of the dye, Congo Red, for biological analyses.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-hydroxy-5-iodobenzoic acid is a reagent in various organic chemical reactions leading to pharmaceutical compounds. It is involved in the synthesis of labeled benzamide analogs for tumor imaging as well as the synthesis of the dye, Congo Red, for biological analyses.

Solubility
Solubility in methanol is almost transparent.

Notes
Light sensitive. Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text