Thermo Scientific Chemicals

2-Nitrobenzhydrazide, 98%, Thermo Scientific Chemicals

Catalog number: B21330.14
25 g, Each
Thermo Scientific Chemicals

2-Nitrobenzhydrazide, 98%, Thermo Scientific Chemicals

Catalog number: B21330.14
25 g, Each
Quantity
Catalog number: B21330.14
also known as B21330-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
606-26-8
IUPAC Name
2-nitrobenzohydrazide
Molecular Formula
C7H7N3O3
InChI Key
LYGGDXLOJMNFBV-UHFFFAOYSA-N
SMILES
NNC(=O)C1=CC=CC=C1[N+]([O-])=O
Appearance (Color)
Cream to yellow to brown
Form
Powder or crystalline powder
Assay (HPLC)
≥97.5%
Melting Point (clear melt)
118.0-125.0?C

Description

The reaction of 2,3-dichloro-1,4-naphthoquinone with p-nitrobenzhydrazide yeilds the disubstituted product bis-(p-nitrobenzhydrazino)-1,4-napthtquinone.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
The reaction of 2,3-dichloro-1,4-naphthoquinone with p-nitrobenzhydrazide yeilds the disubstituted product bis-(p-nitrobenzhydrazino)-1,4-napthtquinone.

Solubility
Soluble in methanol(almost transparency).

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text