4-Aminomorpholine, 95%
4-Aminomorpholine, 95%
4-Aminomorpholine, 95%
4-Aminomorpholine, 95%
Thermo Scientific Chemicals

4-Aminomorpholine, 95%

CAS: 4319-49-7 | C4H10N2O | 102.137 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Catalog number B21381.06
also known as B21381-06
Price (EUR)
66,70
Each
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (EUR)
66,70
Each
Add to cart
Chemical Identifiers
CAS4319-49-7
IUPAC Namemorpholin-4-amine
Molecular FormulaC4H10N2O
InChI KeyMKQLBNJQQZRQJU-UHFFFAOYSA-N
SMILESNN1CCOCC1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)≥97.5%
Refractive Index1.4765-1.4805 @ 20?C
Appearance (Color)Clear colorless
FormLiquid
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only