Thermo Scientific Chemicals

Ammonium iron(III) oxalate trihydrate, 98%, Thermo Scientific Chemicals

Catalog number: B21411.36
500 g, Each
Thermo Scientific Chemicals

Ammonium iron(III) oxalate trihydrate, 98%, Thermo Scientific Chemicals

Catalog number: B21411.36
500 g, Each
Quantity
Catalog number: B21411.36
also known as B21411-36
Price (USD)
Price: 156.00
Online price: 132.65
Your price:
Quantity
-

Chemical Identifiers

CAS
13268-42-3
IUPAC Name
triammonium hexaoxo-1,4,6,9,10,13-hexaoxa-5-ferraspiro[4.4⁵.4⁵]tridecane-5,5,5-tris(ylium) trihydrate
Molecular Formula
C6H18FeN3O15
InChI Key
ARJSTCBPHIPXIJ-UHFFFAOYSA-K
SMILES
[NH4+].[NH4+].[NH4+].O.O.O.O=C1O[Fe+3]234(OC1=O)OC(=O)C(=O)O2.O=C(O3)C(=O)O4
Appearance (Color)
Green
Form
Crystals or powder or crystalline powder or fused solid
Assay (Redox Titration)
≥97.5 to ≤102.5%
Water Content (Karl Fischer Titration)
10.75-14.43% (2.5-3.5 waters)

Description

Ammonium iron(III) oxalate trihydrate is used in the preparation of Langmuir-Blodgett magnetic thin films. It is also involved in the production of blueprint paper.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ammonium iron(III) oxalate trihydrate is used in the preparation of Langmuir-Blodgett magnetic thin films. It is also involved in the production of blueprint paper.

Solubility
Soluble in water. Insoluble in ethanol.

Notes
Light sensitive and hygroscopic. Incompatible with strong oxidizing agents and strong acids.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text