Thermo Scientific Chemicals

Selenourea, 99%, Thermo Scientific Chemicals

Catalog number: B21440.06
5 g, Each
Thermo Scientific Chemicals

Selenourea, 99%, Thermo Scientific Chemicals

Catalog number: B21440.06
5 g, Each
Quantity
Catalog number: B21440.06
also known as B21440-06
Price (USD)
Price: 305.00
Online price: 259.65
Your price:
Quantity
-

Chemical Identifiers

CAS
630-10-4
IUPAC Name
[(C-selanylcarbonimidoyl)amino]-λ¹-selanyl
Molecular Formula
CH3N2Se
InChI Key
ZUCFIOXGLBMWHV-UHFFFAOYSA-N
SMILES
NC([Se])=N
Appearance (Color)
White to grey or red to purple
Assay from Supplier's CofA
Selenium (Se): 63.22-65.15% (titration)
Form
Powder or crystals or crystalline powder
Melting Point
200±10°C

Description

Selenourea is used in the synthesis of selenium heterocycles, which exhibits anti-inflammatory and anti-tumor activity. It acts as an effective ligand and form complexes with transition metals and metalloids.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Selenourea is used in the synthesis of selenium heterocycles, which exhibits anti-inflammatory and anti-tumor activity. It acts as an effective ligand and form complexes with transition metals and metalloids.

Solubility
Soluble in water, alcohol and ether.

Notes
Air and moisture sensitive. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text