L-(+)-Asparagine, 99%
L-(+)-Asparagine, 99%
L-(+)-Asparagine, 99%
L-(+)-Asparagine, 99%
Thermo Scientific Chemicals

L-(+)-Asparagine, 99%

CAS: 70-47-3 | C4H8N2O3 | 132.119 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
2500 g
Catalog number B21473.36
also known as B21473-36
Price (EUR)
287,00
Each
Add to cart
Quantity:
500 g
Request bulk or custom format
Price (EUR)
287,00
Each
Add to cart
Chemical Identifiers
CAS70-47-3
IUPAC Name2-amino-3-carbamoylpropanoic acid
Molecular FormulaC4H8N2O3
InChI KeyDCXYFEDJOCDNAF-UHFFFAOYNA-N
SMILESNC(CC(N)=O)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
CommentPurchased in the U.S. and in other countries
Optical Rotation+35.0° ± 2.0° (c=10 in 2N HCl)
FormPowder or crystalline powder or crystals
Water Content (Karl Fischer Titration)≤2.5%
View more
L-(+)-Asparagine acts as a precursor to aspartate. It is also used as a diuretic. It is also employed in the biosynthesis of glycoproteins and proteins. It acts as a food additive. It reacts with reducing sugar to yield acrylamide, which finds application in french fries, potato chips, and toasted bread. It plays an important role in the function and development of the brain. Further, it is useful for human body, physically and mentally. In addition to this, it is used in cell culture media and is a component of (2-methoxyethoxy)methyl (MEM) non-essential amino acid solution.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
L-(+)-Asparagine acts as a precursor to aspartate. It is also used as a diuretic. It is also employed in the biosynthesis of glycoproteins and proteins. It acts as a food additive. It reacts with reducing sugar to yield acrylamide, which finds application in french fries, potato chips, and toasted bread. It plays an important role in the function and development of the brain. Further, it is useful for human body, physically and mentally. In addition to this, it is used in cell culture media and is a component of (2-methoxyethoxy)methyl (MEM) non-essential amino acid solution.

Solubility
Soluble in acids, bases. Insoluble in methanol, ethanol, ether and benzene.

Notes
Incompatible with strong oxidizing agents.
RUO – Research Use Only

General References:

  1. Zhang, J.; Guo, Y.; Lu, T.; Shi, H.; Dai, T.; Shi, T. Oxidation of L-asparagine by bis(hydrogenperiodato)argentate(III): a kinetic investigation. Transition Met. Chem. 2015, 40 (3), 281-286.
  2. Asghar, B. H.; Altass, H. M.; Fawzy, A. Transition metal-catalyzed oxidation of L-asparagine by platinum(IV) in acid medium: a kinetic and mechanistic study. Trans. Metal Chem. 2015, 40 (6), 587-594.