Thermo Scientific Chemicals

2,5-Dimethylstyrene, 97%, stab. with 4-tert-butylcatechol, Thermo Scientific Chemicals

Catalog number: B21476.14
25 g, Each
Thermo Scientific Chemicals

2,5-Dimethylstyrene, 97%, stab. with 4-tert-butylcatechol, Thermo Scientific Chemicals

Catalog number: B21476.14
25 g, Each
Quantity
Catalog number: B21476.14
also known as B21476-14
Price (TWD)

Chemical Identifiers

CAS
2039-89-6
IUPAC Name
2-ethenyl-1,4-dimethylbenzene
Molecular Formula
C10H12
InChI Key
DBWWINQJTZYDFK-UHFFFAOYSA-N
SMILES
CC1=CC=C(C)C(C=C)=C1
Appearance (Color)
Clear colourless to pale yellow to pale orange
Assay (GC)
≥96.0%
Refractive Index
1.5365-1.5415 @20?C
Solution Test
10% in diethyl ether: clear
Solution Test
10% in methanol: clear

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text