Use Prolong Diamond with any Fluorescent Dye or Fluorescent Protein for brightest images. Learn more.
Citations & References (15)
Invitrogen™
BrdUTP (5-Bromo-2'-Deoxyuridine 5'-Triphosphate), 10 mM in TE buffer
BrdUTP can be used in conjunction with tetrminal deoxynucleotidyl transferase (TdT) to label apoptotic cells in the TUNEL assay. BrdUTPRead more
Have Questions?
Catalog Number
Quantity
B21550
25 μL
Catalog number B21550
Price (USD)
231.57
Special offer
Online exclusive
Ends: 27-Jun-2026
249.00
Save 17.43 (7%)
Each
Quantity:
25 μL
Price (USD)
231.57
Special offer
Online exclusive
Ends: 27-Jun-2026
249.00
Save 17.43 (7%)
Each
BrdUTP can be used in conjunction with tetrminal deoxynucleotidyl transferase (TdT) to label apoptotic cells in the TUNEL assay. BrdUTP can be detected with anti-BrdU antibodies.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Label or DyeOther Label(s) or Dye(s)
Product TypeBrdUTP
Quantity25 μL
Shipping ConditionWet Ice
Concentration10 mM
Unit SizeEach
Contents & Storage
Store in freezer -5°C to -30°C and protect from light.
Frequently asked questions (FAQs)
Can dU-containing DNA produced by amplification with dUTP be cut by restriction enzymes?
You will need to confirm with the manufacturer of the restriction enzyme you are using to find out whether or not it can recognize dUTP-containing DNA.
Immunoseparation and immunodetection of nucleic acids labeled with halogenated nucleotides.
Authors:Haider SR, Juan G, Traganos F, Darzynkiewicz Z
Journal:Exp Cell Res
PubMed ID:9260920
'A novel methodology for labeling, isolation, and detection of nucleic acids is described. Nucleic acid isolation is based on in vivo or in vitro incorporation of BrU or BrdU to either RNA or DNA, respectively, followed by immunoprecipitation of the labeled nucleic acid utilizing anti-BrdU MoAb, which crossreacts with BrU, ... More
Purification and characterization of human topoisomerase I mutants.
Authors:Jensen AD, Svejstrup JQ
Journal:Eur J Biochem
PubMed ID:8612607
'A system for rapid purification and characterization of eukaryotic topoisomerase-I mutants has been developed. The system utilizes six-histidine tagging of human topoisomerase I expressed in Saccharomyces cerevisiae to enable purification by nickel-affinity chromatography. Virtually homogenous mutant proteins are then tested for their ability to relax supercoiled DNA plasmids and their ... More
Flow cytometric evaluation of apoptosis, necrosis and recovery when culturing monocytes.
'After developing and applying a method for cryopreserving monocytes, we found a substantial cell loss when culturing these cells. Monocytes were isolated from blood donors by density gradient centrifugation, purified by elutriation and cryopreserved. Thawed cells were cultured in ultra low attachment wells and studied with Annexin V, Propidium iodide, ... More
A sensitive non-isotopic assay specific for HIV-1 associated reverse transcriptase.
Authors:Porstmann T, Meissner K, Glaser R, Döpel SH, Sydow G
Journal:J Virol Methods
PubMed ID:1713913
'A sensitive non-isotopic assay for specific detection of reverse transcriptase (RT) of the human immunodeficiency virus type 1 (HIV-1) is described using 5-bromo-2''-deoxyuridine triphosphate (BrdUTP) instead of tritiated thymidine triphosphate. After the RT reaction the template primer is degraded by alkaline hydrolysis. Single-stranded poly.(BrdU) is detected in an immunoenzymometric assay ... More
A sensitive assay for the quantification of reverse transcriptase activity based on the use of carrier-bound template and non-radioactive-product detection, with special reference to human-immunodeficiency-virus isolation.
'A non-radioactive 96-well microtitre plate reverse transcriptase (RT) assay, based on the use of covalently bound riboadenosine homopolymer in the wells and 5-bromodeoxyuridined 5''-triphosphate (BrdUTP) as dNTP, is described. The whole assay is performed in a single well, including the quantitative detection of incorporated BrdU, which is performed immunologically using ... More