2,5-Difluorophenylacetic acid, 97%
2,5-Difluorophenylacetic acid, 97%
2,5-Difluorophenylacetic acid, 97%
2,5-Difluorophenylacetic acid, 97%
Thermo Scientific Chemicals

2,5-Difluorophenylacetic acid, 97%

CAS: 85068-27-5 | C8H6F2O2 | 172.131 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
Catalog number B21595.14
also known as B21595-14
Price (EUR)
-
Quantity:
25 g
Request bulk or custom format
Chemical Identifiers
CAS85068-27-5
IUPAC Name2-(2,5-difluorophenyl)acetate
Molecular FormulaC8H5F2O2
InChI KeyFKCRTRYQHZHXES-UHFFFAOYSA-M
SMILES[O-]C(=O)CC1=CC(F)=CC=C1F
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)≥96.0%
Appearance (Color)White to pale cream
FormCrystals or powder or crystalline powder
Assay (Aqueous acid-base Titration)≥96.0 to ≤104.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only