Tetrafluoroisophthalonitrile, 97%
Tetrafluoroisophthalonitrile, 97%
Tetrafluoroisophthalonitrile, 97%
Thermo Scientific Chemicals

Tetrafluoroisophthalonitrile, 97%

CAS: 2377-81-3 | C8F4N2 | 200.10 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
Catalog number B21599.03
also known as B21599-03
Price (TWD)
-
Quantity:
1 g
Request bulk or custom format
Chemical Identifiers
CAS2377-81-3
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to pale cream or pale pink
Identification (FTIR)Conforms
Melting Point71.0-80.0?C
FormCrystals or powder or crystalline powder
Assay (GC)≥96.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only