Thermo Scientific Chemicals

Isopentyl acetate, 99%, cont. ca 10% other isomers, Thermo Scientific Chemicals

Catalog number: B21618.AP
500 mL, Each
Thermo Scientific Chemicals

Isopentyl acetate, 99%, cont. ca 10% other isomers, Thermo Scientific Chemicals

Catalog number: B21618.AP
500 mL, Each
Quantity
Catalog number: B21618.AP
also known as B21618-AP
Price (USD)
Price: 88.90
Online price: 75.65
Your price:
Quantity
-

Chemical Identifiers

CAS
123-92-2
IUPAC Name
3-methylbutyl acetate
Molecular Formula
C7H14O2
InChI Key
MLFHJEHSLIIPHL-UHFFFAOYSA-N
SMILES
CC(C)CCOC(C)=O
Assay (GC)
>98.5% (sum of isomers)
Refractive Index
1.3990 - 1.4035 @20?C
Appearance (Color)
Clear, colourless
Form
liquid

Description

It is used as a masking agent, perfuming agent, and solvent in cosmetics industry. Mainly used in flavors and because of its intensive fruity taste especially for fruit flavors such as banana, strawberry, pear, apple and cherry flavors. Traces are used in certain sweet spicy types such as vanilla flavors. Occasionally used in perfumery when a fresh-fruity note is demanded. The major application is the use in nail polish removers and nail polish as a solvent. Isopentyl acetate is a widely employed ester.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as a masking agent, perfuming agent, and solvent in cosmetics industry. Mainly used in flavors and because of its intensive fruity taste especially for fruit flavors such as banana, strawberry, pear, apple and cherry flavors. Traces are used in certain sweet spicy types such as vanilla flavors. Occasionally used in perfumery when a fresh-fruity note is demanded. The major application is the use in nail polish removers and nail polish as a solvent. Isopentyl acetate is a widely employed ester.

Solubility
Soluble in alcohol, fixed oils, kerosene and paraffin oil. Insoluble in water, glycerin and propylene glycol.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text