2-Amino-6-bromopurine, 98%
2-Amino-6-bromopurine, 98%
2-Amino-6-bromopurine, 98%
2-Amino-6-bromopurine, 98%
Thermo Scientific Chemicals

2-Amino-6-bromopurine, 98%

CAS: 82499-03-4 | C5H4BrN5 | 214.026 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number B21629.06
also known as B21629-06
Price (USD)
139.65
Online Exclusive
155.00
Save 15.35 (10%)
Each
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)
139.65
Online Exclusive
155.00
Save 15.35 (10%)
Each
Add to cart
Chemical Identifiers
CAS82499-03-4
IUPAC Name6-bromo-7H-purin-2-amine
Molecular FormulaC5H4BrN5
InChI KeyHPGBGNVPUMCKPM-UHFFFAOYSA-N
SMILESNC1=NC(Br)=C2NC=NC2=N1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Yellow to orange
Assay (Non-aqueous acid-base Titration)≥97.5 to ≤102.5%
Assay (HPLC)≥97.5%
FormPowder
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only