Thermo Scientific Chemicals

Trifluoromethanesulfonyl chloride, 98%, Thermo Scientific Chemicals

Catalog number: B21774.22
100 g, Each
Thermo Scientific Chemicals

Trifluoromethanesulfonyl chloride, 98%, Thermo Scientific Chemicals

Catalog number: B21774.22
100 g, Each
Quantity
Catalog number: B21774.22
also known as B21774-22
Price (USD)
Price: 925.00
Online price: 808.65
Your price:
Quantity
-

Chemical Identifiers

CAS
421-83-0
Appearance (Color)
Clear or slightly turbid, colorless to yellow
Assay (GC)
>97.5%
Refractive Index
1.3310-1.3370 @ 20?C
Form
Liquid

Description

Ru(II) complex-catalyzed trifluoromethylating agent for aromatics and alkenes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ru(II) complex-catalyzed trifluoromethylating agent for aromatics and alkenes.

Solubility
Hydrolyzes with water.

Notes
Moisture sensitive. Store away from water/moisture, heat and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text