Thermo Scientific Chemicals

Digoxin, 96%, Thermo Scientific Chemicals

Catalog number: B21902.06
5 g, Each
Thermo Scientific Chemicals

Digoxin, 96%, Thermo Scientific Chemicals

Catalog number: B21902.06
5 g, Each
Quantity
Catalog number: B21902.06
also known as B21902-06
Price (USD)
Price: 465.00
Online price: 405.65
Your price:
Quantity
-

Chemical Identifiers

CAS
20830-75-5
IUPAC Name
4-[(1R,3aS,3bR,5aR,7S,9aS,9bS,11R,11aS)-7-{[(2R,4S,5S,6R)-5-{[(2S,4S,5S,6R)-5-{[(2S,4S,5S,6R)-4,5-dihydroxy-6-methyloxan-2-yl]oxy}-4-hydroxy-6-methyloxan-2-yl]oxy}-4-hydroxy-6-methyloxan-2-yl]oxy}-3a,11-dihydroxy-9a,11a-dimethyl-hexadecahydro-1H-cyclopenta[a]phenanthren-1-yl]-2,5-dihydrofuran-2-one
Molecular Formula
C41H64O14
InChI Key
LTMHDMANZUZIPE-PUGKRICDSA-N
SMILES
C[C@H]1O[C@H](C[C@H](O)[C@@H]1O)O[C@H]1[C@@H](O)C[C@H](O[C@H]2[C@@H](O)C[C@H](O[C@H]3CC[C@@]4(C)[C@H](CC[C@@H]5[C@@H]4C[C@@H](O)[C@]4(C)[C@H](CC[C@]54O)C4=CC(=O)OC4)C3)O[C@@H]2C)O[C@@H]1C
Assay (HPLC)
≥95.0%
Optical Rotation
12.0 ± 0.1? (C=10 in pyridine)
Form
Powder
Appearance (Color)
White

Description

Digoxin is used in several cardiac pathologies such as atrial fibrillation and heart failure.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Digoxin is used in several cardiac pathologies such as atrial fibrillation and heart failure.

Solubility
Soluble in dimethyl sulfoxide and methanol.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text