Ammonium iron(III) hexacyanoferrate(II) hydrate, tech.
Ammonium iron(III) hexacyanoferrate(II) hydrate, tech.
Ammonium iron(III) hexacyanoferrate(II) hydrate, tech.
Ammonium iron(III) hexacyanoferrate(II) hydrate, tech.
Thermo Scientific Chemicals

Ammonium iron(III) hexacyanoferrate(II) hydrate, tech.

CAS: 25869-00-5 | C6H4Fe2N7 | 285.837 g/mol
Have Questions?
變更視圖buttonViewtableView
Quantity:
25 g
100 g
產品號碼 B21908.22
亦稱為 B21908-22
價格 (TWD)
-
申請報價
Quantity:
100 g
Request bulk or custom format
化學識別
CAS1246969-06-1
規格Specification Sheet規格表
Appearance (Color)Dark blue
FormPowder
Loss on Drying≤12% @105°C
Solution TestInsoluble in water
It is employed as a intermediate for pharmaceuticals. Also used in decorative cosmetics.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is employed as a intermediate for pharmaceuticals. Also used in decorative cosmetics.

Solubility
Soluble in water and mineral acids. Insoluble in alkali.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
RUO – Research Use Only

General References:

  1. Kirin Escher; Frank G. Shellock. Evaluation of MRI artifacts at 3 Tesla for 38 commonly used cosmetics. Magnetic Resonance Imaging, 2013, 31 (5), 778-782.
  2. H. H. Van Tuyl; R. L Moore. Recovery of Fission Product Cesium from Acidic Wastes. Ind. Eng. Chem, 1959, 51 (6), 741-744.