Trimethyl 1,3,5-benzenetricarboxylate, 98%
Trimethyl 1,3,5-benzenetricarboxylate, 98%
Trimethyl 1,3,5-benzenetricarboxylate, 98%
Thermo Scientific Chemicals

Trimethyl 1,3,5-benzenetricarboxylate, 98%

CAS: 2672-58-4 | C12H12O6 | 252.222 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number B21926.14
also known as B21926-14
Price (EUR)
121,00
Each
Quantity:
25 g
Request bulk or custom format
Price (EUR)
121,00
Each
Chemical Identifiers
CAS2672-58-4
IUPAC Name1,3,5-trimethyl benzene-1,3,5-tricarboxylate
Molecular FormulaC12H12O6
InChI KeyRGCHNYAILFZUPL-UHFFFAOYSA-N
SMILESCOC(=O)C1=CC(=CC(=C1)C(=O)OC)C(=O)OC
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Assay (GC)≥97.5%
Identification (FTIR)Conforms
Melting Point (clear melt)141-147?C
FormCrystals or powder or crystalline powder
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only