Thermo Scientific Chemicals

Di-n-octadecyl disulfide, 98%, Thermo Scientific Chemicals

Catalog number: B21978.14
25 g, Each
Thermo Scientific Chemicals

Di-n-octadecyl disulfide, 98%, Thermo Scientific Chemicals

Catalog number: B21978.14
25 g, Each
Quantity
Catalog number: B21978.14
also known as B21978-14
Price (USD)
Price: 454.00
Online price: 392.65
Your price:
Quantity
-

Chemical Identifiers

CAS
2500-88-1
IUPAC Name
1-(octadecyldisulfanyl)octadecane
Molecular Formula
C36H74S2
InChI Key
MQQKTNDBASEZSD-UHFFFAOYSA-N
SMILES
CCCCCCCCCCCCCCCCCCSSCCCCCCCCCCCCCCCCCC
Appearance (Color)
White
Melting Point
60-65°C
Form
Crystalline powder or powder
Assay from Suppliers CofA
≥97.5% (HPLC)

Description

It is used as a co stabilizer, antioxidant for plastics, polyoletins, HDPE, LDPE, PMMA, PA, engineering thermoplastic elastomers, synergist, thermal stabilizer, processing stabilizer in high temperature applications, peroxide decomposer.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as a co stabilizer, antioxidant for plastics, polyoletins, HDPE, LDPE, PMMA, PA, engineering thermoplastic elastomers, synergist, thermal stabilizer, processing stabilizer in high temperature applications, peroxide decomposer.

Solubility
Insoluble in water.

Notes
Store in cool, dry place in tightly closed containers. Store away from strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text