Hexakis(bromomethyl)benzene, 98%
Hexakis(bromomethyl)benzene, 98%
Hexakis(bromomethyl)benzene, 98%
Thermo Scientific Chemicals

Hexakis(bromomethyl)benzene, 98%

CAS: 3095-73-6 | C12H12Br6 | 635.652 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number B22016.03
also known as B22016-03
Price (EUR)
38,00
Each
Quantity:
1 g
Request bulk or custom format
Price (EUR)
38,00
Each
Chemical Identifiers
CAS3095-73-6
IUPAC Namehexakis(bromomethyl)benzene
Molecular FormulaC12H12Br6
InChI KeyXJOUCILNLRXRTF-UHFFFAOYSA-N
SMILESBrCC1=C(CBr)C(CBr)=C(CBr)C(CBr)=C1CBr
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to pale cream to pale brown
FormPowder
Assay (HPLC)≥97.5%
Melting Point (DSC)290.0-297.0?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only