Tricarballylic acid, 98%
Tricarballylic acid, 98%
Tricarballylic acid, 98%
Thermo Scientific Chemicals

Tricarballylic acid, 98%

CAS: 99-14-9 | C6H8O6 | 176.12 g/mol
Have Questions?
變更視圖buttonViewtableView
Quantity:
5 g
25 g
產品號碼 B22103.06
亦稱為 B22103-06
價格 (TWD)
-
Quantity:
5 g
Request bulk or custom format
化學識別
CAS99-14-9
IUPAC Namepropane-1,2,3-tricarboxylic acid
Molecular FormulaC6H8O6
InChI KeyKQTIIICEAUMSDG-UHFFFAOYSA-N
SMILESOC(=O)CC(CC(O)=O)C(O)=O
檢視更多
規格Specification Sheet規格表
Appearance (Color)White to cream to pale brown
FormCrystals or powder or crystalline powder
Assay (Aqueous acid-base Titration)≥97.5 to ≤102.5%
Melting Point (clear melt)154-160?C
It is used as an alternative to DMDHEU (formaldehyde releasing cross- linking agent).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as an alternative to DMDHEU (formaldehyde releasing cross- linking agent).

Solubility
Soluble in water (500 mg/ml at 18°C), methanol, and ether (slightly).

Notes
Store in cool, dry place in tightly closed containers. Store away from strong oxidizing agents.
RUO – Research Use Only

General References:

  1. Craig D Boyle; Yoshito Kishi. Absolute configuration at the tricarballylic acid moieties of fumonisin B2. Tetrahedron Letters. 1995, 36 (26), 4579-4582.
  2. Frank C. Magne; Robert R. Mod. Plasticizers from Aconitic and Tricarballylic Acids. Ind. Eng. Chem. 1953, 45 (7), 1546-1547.