Thermo Scientific Chemicals

3-Fluorocatechol, 99%, Thermo Scientific Chemicals

Catalog number: B22145.06
5 g, Each
Thermo Scientific Chemicals

3-Fluorocatechol, 99%, Thermo Scientific Chemicals

Catalog number: B22145.06
5 g, Each
Quantity
Catalog number: B22145.06
also known as B22145-06
Price (USD)
Price: 158.00
Online price: 135.65
Your price:
Quantity
-

Chemical Identifiers

CAS
363-52-0
IUPAC Name
3-fluorobenzene-1,2-diol
Molecular Formula
C6H5FO2
InChI Key
DXOSJQLIRGXWCF-UHFFFAOYSA-N
SMILES
OC1=CC=CC(F)=C1O
Appearance (Color)
Cream to pale brown to grey
Form
Crystals or powder or crystalline powder or lumps
Assay (GC)
≥98.5%
Melting Point (clear melt)
65.0-74.0?C

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text