L-Glutamic acid 5-benzyl ester, 99%
L-Glutamic acid 5-benzyl ester, 99%
L-Glutamic acid 5-benzyl ester, 99%
Thermo Scientific Chemicals

L-Glutamic acid 5-benzyl ester, 99%

CAS: 1676-73-9 | C12H15NO4 | 237.255 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number B22184.14
also known as B22184-14
Price (EUR)
219,00
Each
Quantity:
25 g
Request bulk or custom format
Price (EUR)
219,00
Each
Chemical Identifiers
CAS1676-73-9
IUPAC Name2-amino-5-(benzyloxy)-5-oxopentanoic acid
Molecular FormulaC12H15NO4
InChI KeyBGGHCRNCRWQABU-UHFFFAOYNA-N
SMILESNC(CCC(=O)OCC1=CC=CC=C1)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Identification (FTIR)Conforms
Assay (Non-aqueous acid-base Titration)≥98.5 to ≤101.5%
FormCrystals or powder or crystalline powder
Optical Rotation19.0 ± 2.0? (C=1 in acetic acid)
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only